missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroselinic acid, MedChemExpress
Description
Petroselinic acid, a positional isomer of oleic acid, is isolated from the vegetable oil of Coriandrum sativum fruits.
Specifications
Specifications
| Chemical Name or Material | Petroselinic acid |
| Physical Form | Liquid |
| CAS | 593-39-5 |
| Color | Colorless |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H34O2 |
| Solubility Information | DMSO : ≥ 100 mg/mL (354.03 mM) |
| SMILES | CCCCCCCCCCC/C=C\CCCCC(O)=O |
| Molecular Weight (g/mol) | 282.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?