missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum spirit, 40 - 60°C, AR™, Macron Fine Chemicals™
Specifications
Specifications
| CAS | 64742-49-0 |
| Quantity | 2.5 L |
| MDL Number | MFCD00081849 |
| UN Number | 1268 |
| Synonym | Petroleum ether, Gasoline, Petrol |
| IUPAC Name | Petroleum ether |
Safety and Handling
Product Identifier
- Petroleum Ether 30/60. Low boiling point naphta(Benzene <0.1%).
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- MIXTURE LIST-Contain: Petroleum Ether
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?