missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Hydrocarbon oil index concentration (as RIVM oil reference standard): max. 0.5 mg/L
- Total peaks between n-decane (C10H22) & n-tetracontane (C40H82): max. 0.5 mg/L
Specifications
Specifications
| CAS | 64742-49-0 |
| Color | Max. 10 (APHA) |
| Quantity | 2.5 L |
| Residue after Evaporation | max. 5 ppm |
| MDL Number | MFCD00081849 |
| UN Number | 1268 |
| Water | max. 0.05% |
| Synonym | Petroleum ether, Gasoline, Petrol |
| IUPAC Name | Petroleum ether |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?