missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, TRC

Product Code. 30421484
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421484 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 30421484 Supplier Toronto Research Chemicals Supplier No. TRCP29350550ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material UnleadedSuperGasolinePlus(unweathered)
CAS 8032-32-4
Quantity 50 mL
Synonym Ligroin, Painters' naphtha, Refined solvent naphtha, Skellysolve F, Skellysolve G, Varnish makers' and painters' naphtha, Benzoline, Canadol
SMILES O=C(C(F)(F)F)N(C)[Si](C)(C)C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.