missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, Spectrophotometric Grade, Solstice

Product Code. 15662800
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15662800 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15662800 Supplier Honeywell Riedel-de Haën Supplier No. 2617341L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Specifications

CAS 8032-32-4
Density 0.64g/mL at 25°C
Quantity 1 L
Explosion Limit 0.08
Flash Point −49°C (−56.2°F)
Ignition Point 475°F
Packaging Glass Bottle
Vapor Pressure 25.8psi (55°C), 7.99psi (20°C)
MDL Number MFCD00081849
UN Number UN1268
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide
Boiling Point 40°C to 60°C
Grade Spectrophotometry
Chemical Name or Material Petroleum Ether
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hydrocarbons, C6, pentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.