missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, puriss. p.a., ACS Reagent, Reag. ISO, Solstice
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Appearance | Liquid |
| Density | 0.640 to 0.655g/mL at 20°C |
| Quantity | 4 x 5 L |
| Flash Point | −30°C (−22°F) |
| Packaging | Aluminum Bottle |
| Residue after Evaporation | 0 to 0.001% |
| Titratable Acid | Free acid (as CH3COOH): ≤0.002% |
| UN Number | UN1268 |
| Boiling Point | 40°C to 60°C |
| Grade | ACS/puriss. p.a. |
| Show More |
Safety and Handling
Product Identifier
- Petroleum ether
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: Hydrocarbons, C6, Isoalkanes, <5% n-Hexane
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?