missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum ether, pure, boiling range ca. 180-280°C, Thermo Scientific Chemicals
59.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Metal drum
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 0.7900g/mL |
Boiling Point | 200.0°C to 250.0°C |
Flash Point | 82°C |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Refractive Index | 1.4300 to 1.4500 |
Quantity | 2.5 L |
Merck Index | 15,5342 |
Specific Gravity | 0.79 |
Show More |
Safety and Handling
Product Identifier
- Petroleum ether, boiling range ca. 180-280°C
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H304-May be fatal if swallowed and enters airways.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'