Product Code. 16626153
Click to view available options
Packaging:
Aluminum bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
16626153
5 L
Aluminum bottle
5 L
15611530
4 x 5 L
Aluminum bottle
Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
16626153
Supplier
Fluka
Supplier No.
159675L
£133.00 £110.95 / 5 L Save £22.05 16% Off
Estimated Shipment: 18-12-2026Log in to see stock.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Laboratory Reagent, bp 40-60 °C
≥95% for general laboratory use.
Chemical Identifiers
CAS
64742-49-0
Molecular Formula
C6H14
Molecular Weight (g/mol)
86.178
MDL Number
MFCD00081849
InChI Key
AFABGHUZZDYHJO-UHFFFAOYSA-N
PubChem CID
7892
IUPAC Name
2-methylpentane
SMILES
CCCC(C)C
Specifications
Color
Colorless
Density
0.65g/cu.cm at 20°C
Boiling Point
40°C to 60°C (1013 hPa)
Flash Point
−30°C (−22°F)
Odor
Characteristic
Packaging
Aluminum bottle
Quantity
5 L
UN Number
UN1268
Synonym
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane
Solubility Information
Practically insoluble in water
Grade
Laboratory Reagent
Physical Form
Liquid
Chemical Name or Material
Petroleum Ether
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?