Product Code. 15524055
Shop All J.T.Baker Products
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
15524055
2.5 L
2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
15524055
Supplier
JT Baker
Supplier No.
5280.2500
Estimated Shipment: 30-12-2026Log in to see stock.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Solvents specially developed for use in evaluation of pesticide and insecticide residue in food, feed, water or soil samples.
Solvents specially developed for use in evaluation of pesticide residue in food, feed, water or soil samples. They are specified to the required low residue levels (5 ppm) for use in research and quality control laboratories.
Specifications
Melting Point
-40°C
CAS
8032-32-4
Color
Colorless
Acidity
max. 0.0005 meq/g
Quantity
2.5 L
Explosion Limit
0.6 to 8 vol%
Flash Point
-18°C
Ignition Point
250°C
Odor
Characteristic
Packaging
Glass
Physical Form
Liquid
Residue after Evaporation
max. 5 ppm
UN Number
1268
Water
max. 0.02 %
Synonym
p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide
Boiling Point
40°C to 60°C
Solubility Information
Miscibility with water at 20 °C is 0.020 g/l
Grade
Pesticide
Chemical Name or Material
Petroleum Ether, Low boiling point naphta (Benzene <0.1%)
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?