Product Code. 12616757
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic Bottle
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
unitSize
12616757
1 L
Amber Glass Bottle
1 L
11408722
500 mL
Amber Glass Bottle
500mL
10254200
2.5 L
Amber Glass Bottle
2.5 L
10204062
2.5 L
Plastic-coated Glass Bottle
2.5 L
10171480
5 L
Plastic Bottle
5 L
10589380
25 L
Metal Can
25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
12616757
Supplier
Fisher Chemical
Supplier No.
P/1440/15
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Petroleum Ether 40-60°C, Extra Pure, SLR
Chemical Identifiers
CAS
64742-49-0
Molecular Formula
C6H14
Molecular Weight (g/mol)
86.178
MDL Number
MFCD00081849
InChI Key
AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID
7892
SMILES
CCCC(C)C
Specifications
Color
Colorless
Quantity
1 L
Packaging
Amber Glass Bottle
Physical Form
Liquid
Vapor Pressure
38kPa at 20°C
Viscosity
0.45 mm2/s at 25°C (ASTM D445)
Boiling Point
40°C
IUPAC Name
Petroleum ether
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?