missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether 40-60°C, Extra Pure, SLR, Fisher Chemical™

Product Code. 10589380 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic Bottle
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
500mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10589380

Brand: Fisher Chemical P/1440/25

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Petroleum Ether 40-60°C, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
SMILES CCCC(C)C

Specifications

Color Colorless
Boiling Point 40°C
Quantity 25 L
Packaging Metal Can
Physical Form Liquid
Vapor Pressure 38kPa at 20°C
Viscosity 0.45 mm2/s at 25°C (ASTM D445)
IUPAC Name 2-methylpentane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.