missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether 40-60°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™

Product Code. 10204350 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Fluorinated HDPE Bottle
Metal Can
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10204350

Brand: Fisher Chemical P/1760/25

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

C6H14, CAS Number-64742-49-0, 1,1-dimethylbutane, isohexane, 2-methylpentan, dimethylpropylmethane, 2-methyl-pentane, 2-methyl pentane, unii-49ib0u6mld, pentane, 2-methyl, methyl pentane, iso-hexane, 2.5L, 40 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
PubChem CID 7892
SMILES CCCC(C)C

Specifications

Color Colorless
Boiling Point 40°C
Quantity 25 L
Packaging Metal Can
Physical Form Liquid
Vapor Pressure 38kPa at 20°C
Viscosity 0.45 mm2/s at 25°C (ASTM D445)
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
IUPAC Name 2-methylpentane
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether, boiling range 40-60°C
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.