missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether 40-60°C, Certified AR for Analysis, Low In Aromatics, Fisher Chemical

Product Code. 10696262 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10696262 2.5 L Amber Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10696262 Supplier Fisher Chemical Supplier No. P/2000/17
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Specifications

CAS 64742-49-0
Color Colorless
Boiling Point 40°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Molecular Formula C6H14
MDL Number MFCD00081849
Vapor Pressure 38kPa at 20°C
Viscosity 0.45 mm2/s at 25°C (ASTM D445)
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
SMILES CCCC(C)C
IUPAC Name Petroleum ether
Molecular Weight (g/mol) 86.178
PubChem CID 7892
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.