missing translation for 'onlineSavingsMsg'
Learn More

Solstice Petroleum Ether

Product Code. 16686153
Change view
Click to view available options
Quantity:
18 L
200 L
Unit Size:
18 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16686153 200 L 200 L
15954750 18 L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 16686153 Supplier Solstice Supplier No. 24587200L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Petroleum Ether, Puriss., low boiling point hydrogen treated naphtha, meets analytical specification of DAB, 40°C to 60°C (bp min. 75%).

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals.
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 64742-49-0
MDL Number MFCD00081849
Synonym Petroleum benzin
IUPAC Name Petroleum ether

Specifications

pH Not applicable
Melting Point No data available
CAS Min % ≥50.00%
Color Colorless
CAS Max % ≤100.00%
Density 0.65 g/cm3 (20°C)
Quantity 200 L
Flash Point -30°C
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line Puriss.
Vapor Pressure No data available
Viscosity No data available
UN Number UN3295
Boiling Point 40°C to 60°C (1.013 hPa)
Percent Purity ≥75.00%
Grade Pharmacopeia
Chemical Name or Material Petroleum ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.