missing translation for 'onlineSavingsMsg'
Learn More

Solstice Petroleum Ether

Product Code. 15954750
Change view
Click to view available options
Quantity:
18 L
200 L
Unit Size:
18 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15954750 18 L 18 L
16686153 200 L 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15954750 Supplier Solstice Supplier No. 2458718L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Petroleum Ether, Puriss., low boiling point hydrogen treated naphtha, meets analytical specification of DAB, 40°C to 60°C (bp min. 75%).

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals.
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 64742-49-0
MDL Number MFCD00081849
Synonym Petroleum benzin
IUPAC Name Petroleum ether

Specifications

pH Not applicable
Melting Point No data available
CAS Min % ≥50.00%
Color Colorless
CAS Max % ≤100.00%
Density 0.65 g/cm3 (20°C)
Quantity 18 L
Flash Point -30°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss.
Vapor Pressure No data available
Viscosity No data available
UN Number UN3295
Boiling Point 40°C to 60°C (1.013 hPa)
Percent Purity ≥75.00%
Grade Pharmacopeia
Chemical Name or Material Petroleum ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.