missing translation for 'onlineSavingsMsg'
Learn More

Solstice Petroleum Ether

Product Code. 15915620
Change view
Click to view available options
Quantity:
45 L
Unit Size:
45 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15915620 45 L 45 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15915620 Supplier Solstice Supplier No. 3229945L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Petroleum Ether, Puriss. p.a., ACS Reagent, Reag. ISO, low boiling point hydrogen treated naphtha, 40°C to 60°C (bp ≥ 90%).

  • ACS general use solvents.
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
  • Suitable for less demanding applications such as routine isocratic HPLC.
  • Variations and decrease issues traceable to solvent contamination.
  • All solvents are tested to meet ACS general use specifications.

Specifications

pH Not applicable
Melting Point No data available
CAS 64742-49-0
CAS Min % ≥50.00%
Color Colorless
CAS Max % ≤100.00%
Density 0.65 g/cm3 (20°C)
Quantity 45 L
Flash Point -30°C
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line Puriss. p.a.
Vapor Pressure No data available
MDL Number MFCD00081849
Viscosity No data available
UN Number UN3295
Synonym Petroleum benzin
Boiling Point 40°C to 60°C (1.013 hPa)
IUPAC Name Petroleum ether
Percent Purity ≥90.00%
Grade ACS
Chemical Name or Material Petroleum ether
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hydrocarbons, C6, Isoalkanes, <5% n-Hexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.