missing translation for 'onlineSavingsMsg'
Learn More

Radleys™ PetriSEAL™ White Sealing Tape

Product Code. 12348049
Encompass
Click to view available options
Color:
White
Unit Size:
1 roll
This item is not returnable. View return policy

Product Code. 12348049

Brand: Radleys™ SEL560030L

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
The unique elastomer characteristics of PetriSEAL™ allows the tape to stretch and conform around the irregular perimeter of these vessels to form a tight seal. Blocks water vapour loss, reduces the rate of gas exchange and prevents the entry of contaminants. Waterproof and chemically resistant to most organic solvents and caustic agents. Flexible down to -30°C and will withstand continuous exposure to temperatures from -40°C to +77°C (with intermittent exposure up to 100°C). 33m roll. Designed for the requirements of sealing the lids of Petri dishes, microtitre plates and culture plates.

Specifications

Applications Sealing the lids of Petri dishes, microplates and culture plates
Color White
Product Type PetriSEAL™ White Sealing Tape
Length (Metric) 33 m
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.