missing translation for 'onlineSavingsMsg'
Learn More

Chroma Gesellschaft™ Petri Seal Tape

Product Code. 12338049
Encompass_Preferred
Change view
Click to view available options
Color:
Clear
Unit Size:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12338049 Clear 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12338049 Supplier Chroma Gesellschaft™ Supplier No. SEL560010R

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
The unique elastomer characteristics of PetriSEAL™ allows the tape to stretch and conform around the irregular perimeter of these vessels to form a tight seal. Blocks water vapour loss, reduces the rate of gas exchange and prevents the entry of contaminants. Waterproof and chemically resistant to most organic solvents and caustic agents. Flexible down to -30°C and will withstand continuous exposure to temperatures from -40°C to +77°C (with intermittent exposure up to 100°C). 33m roll. Designed for the requirements of sealing the lids of Petri dishes, microtitre plates and culture plates.

Specifications

Applications Stretches to conform around the irregular perimeter of vessels to form a tight seal, blocking water vapour loss, reducing the rate of gas exchange and preventing the entry of contaminants
Color Clear
Length (Metric) 33 m
For Use With (Equipment) Petri dishes, microplates and culture plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.