missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Petri Dish Platinum Electrode Chamber

Product Code. p-9571005
Click to view available options
Size (Metric) Gap:
5 mm
15 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15417340

Brand: BTX™ 450506

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Petri Dish Platinum Electrode Chamber for ex vivo tissues gene or drug delivery.

  • Simple and easy to use
  • Available in 5 and 15 mm
  • Reusable, lab-grade Pyrex glass

Specifications

For Use With (Application) For ex Vivo Tissues Gene or Drug Delivery Using Large or Uniquely Shaped Tissue Samples
For Use With (Equipment) ECM™ 830, ECM™ 2001+, Gemini X2, Legacy ECM™ 2001
Material Glass, Platinum
Product Line BTX Electroporation
Product Type Electroporation accessory
Description Petri Dish Platinum Electrode Chamber, 15 mm gap
Depth (English) 0.59 in.
Depth (Metric) 15 mm
Includes Petri Dish Electrode Chamber Only
Length (English) 0.39 in.
Length (Metric) 10 mm
Quantity 1 Ea.
Size (English) Gap 0.59 in.
Size (Metric) Gap 15 mm
Width (English) 0.2 in.
Width (Metric) 5 mm
Sterility Non-sterile
Connection Type Banana Plug to Micrograbber
Pulse Form Square Wave
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.