missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Petri Dish Platinum Electrode Chamber
Click to view available options
Size (Metric) Gap:
5 mm
15 mm
Unit Size:
Each
Description
- Simple and easy to use
- Available in 5 and 15 mm
- Reusable, lab-grade Pyrex glass
Specifications
Specifications
| For Use With (Application) | For ex Vivo Tissues Gene or Drug Delivery Using Large or Uniquely Shapped Tissue Samples |
| For Use With (Equipment) | ECM™ 830, ECM™ 2001+, Gemini X2, Legacy ECM™ 2001 |
| Material | Glass, Platinum |
| Product Line | BTX Electroporation |
| Product Type | Electroporation accessory |
| Description | Petri Dish Platinum Electrode Chamber, 5 mm gap |
| Depth (English) | 0.12 in. |
| Depth (Metric) | 3 mm |
| Includes | Petri Dish Electrode Chamber Only |
| Length (English) | 0.31 in. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction