missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pet1 Antibody (FEV/7311), Novus Biologicals™
Mouse Monoclonal Antibody
£187.00 - £377.00
Specifications
| Antigen | Pet1 |
|---|---|
| Clone | FEV/7311 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694964
|
Novus Biologicals
NBP3-23927-100ug |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629583
|
Novus Biologicals
NBP3-23927-20ug |
20 μg |
£187.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pet1 Monoclonal antibody specifically detects Pet1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Pet1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| 54738 | |
| IgG1 κ | |
| Protein A or G purified |
| FEV/7311 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FEV (ETS oncogene family), FEV (fifth Ewing variant), Fifth Ewing variant protein, HSRNAFEV, PC12 ETS domain-containing transcription factor 1, PC12 ETS factor 1, Pet-1, PET1, protein FEV | |
| Recombinant fragment (around aa1-200) of human Pet1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title