missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Pestle

Pestle

Brand:  Haldenwanger™ 56/3/glazed

Code : 02

Additional Details : Weight : 0.21000kg

 View more versions of this product

Product Code. 10744585

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Grinding surfaces are unglazed.
Specifications

Specifications

42mm
Pestle
175mm
Pestle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers