missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Mixture I 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30328630
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328630 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328630 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000143AL
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-Indoxacarb∣Flufenacet∣Flusilazole∣Flutolanil∣Flutriafol∣Folpet∣Furilazole∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexaconazole∣Imazamethabenz-methyl∣Ipconazole∣Isoxadifen-ethyl∣Kresoxim-methyl∣Lactofen∣Malathion∣Mefenpyr-diethyl∣Metalaxyl∣Metalaxyl-M∣Metconazole
CAS 173584-44-6, 142459-58-3, 85509-19-9, 66332-96-5, 76674-21-0, 133-07-3
Analyte or Component Names (S)-Indoxacarb∣Flufenacet∣Flusilazole∣Flutolanil∣Flutriafol∣Folpet∣Furilazole∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexaconazole∣Imazamethabenz-methyl∣Ipconazole∣Isoxadifen-ethyl∣Kresoxim-methyl∣Lactofen∣Malathion∣Mefenpyr-diethyl∣Metalaxyl∣Metalaxyl-M∣Metconazole
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) (S)-Indoxacarb 100 μg/mL∣Flufenacet 100 μg/mL∣Flusilazole 100 μg/mL∣Flutolanil 100 μg/mL∣Flutriafol 100 μg/mL∣Folpet 100 μg/mL∣Furilazole 100 μg/mL∣Heptachlor 100 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 100 μg/mL∣Hexaconazole 100 μg/mL∣Imazamethabenz-methyl 100 μg/mL∣Ipconazole 100 μg/mL∣Isoxadifen-ethyl 100 μg/mL∣Kresoxim-methyl 100 μg/mL∣Lactofen 100 μg/mL∣Malathion 100 μg/mL∣Mefenpyr-diethyl 100 μg/mL∣Metalaxyl 100 μg/mL∣Metalaxyl-M 100 μg/mL∣Metconazole 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.