missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Mixture for HJ 768-2015 200 μg/mL in Hexane, Dr. Ehrenstorfer

Product Code. 30324880
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324880 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324880 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000586HE
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorpyrifos∣Diazinon∣Dimethoate∣Disulfoton∣Ethion∣Iprobenfos∣Malathion∣Parathion-ethyl∣Parathion-methyl∣Phenthoate∣Phorate∣Profenofos
CAS 2921-88-2, 333-41-5, 60-51-5, 298-04-4, 563-12-2, 26087-47-8
Analyte or Component Names Chlorpyrifos∣Diazinon∣Dimethoate∣Disulfoton∣Ethion∣Iprobenfos∣Malathion∣Parathion-ethyl∣Parathion-methyl∣Phenthoate∣Phorate∣Profenofos
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Hexane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Chlorpyrifos 200 μg/mL∣Diazinon 200 μg/mL∣Dimethoate 200 μg/mL∣Disulfoton 200 μg/mL∣Ethion 200 μg/mL∣Iprobenfos 200 μg/mL∣Malathion 200 μg/mL∣Parathion-ethyl 200 μg/mL∣Parathion-methyl 200 μg/mL∣Phenthoate 200 μg/mL∣Phorate 200 μg/mL∣Profenofos 200 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.