missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Mixture 679 100 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30316570
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316570 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316570 Supplier Dr. Ehrenstorfer Supplier No. DREA50000679AC
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azoxystrobin∣Mepanipyrim∣Myclobutanil∣Pyrimethanil
CAS 131860-33-8, 110235-47-7, 88671-89-0, 53112-28-0
Analyte or Component Names Azoxystrobin∣Mepanipyrim∣Myclobutanil∣Pyrimethanil
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Azoxystrobin 100 μg/mL∣Mepanipyrim 100 μg/mL∣Myclobutanil 100 μg/mL∣Pyrimethanil 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.