missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Mixture 250 100 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30327573
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327573 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327573 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000250AC
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bifenthrin∣Etoxazole∣Fenamidone∣Fenamiphos-sulfone∣Fenamiphos-sulfoxide∣Fenbuconazole∣Fenthion∣lambda-Cyhalothrin∣Phosmet∣Resmethrin∣tau-Fluvalinate∣Tetramethrin∣Triticonazole
CAS 82657-04-3, 153233-91-1, 161326-34-7, 31972-44-8, 31972-43-7, 114369-43-6
Analyte or Component Names Bifenthrin∣Etoxazole∣Fenamidone∣Fenamiphos-sulfone∣Fenamiphos-sulfoxide∣Fenbuconazole∣Fenthion∣lambda-Cyhalothrin∣Phosmet∣Resmethrin∣tau-Fluvalinate∣Tetramethrin∣Triticonazole
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) Bifenthrin 100 μg/mL∣Etoxazole 100 μg/mL∣Fenamidone 100 μg/mL∣Fenamiphos-sulfone 100 μg/mL∣Fenamiphos-sulfoxide 100 μg/mL∣Fenbuconazole 100 μg/mL∣Fenthion 100 μg/mL∣lambda-Cyhalothrin 100 μg/mL∣Phosmet 100 μg/mL∣Resmethrin 100 μg/mL∣tau-Fluvalinate 100 μg/mL∣Tetramethrin 100 μg/mL∣Triticonazole 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.