missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pesticide Mixture 200 100 μg/mL in Ethyl acetate, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Methoxychlor∣Mirex∣oxy-Chlordane∣Quintozene∣Trans-Chlordane (Gamma Isomer) |
| CAS | 53-19-0, 3424-82-6, 789-02-6, 72-54-8, 72-55-9, 50-29-3 |
| Analyte or Component Names | 2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Methoxychlor∣Mirex∣oxy-Chlordane∣Quintozene∣Trans-Chlordane (Gamma Isomer) |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Ethyl Acetate |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 2,4'-DDD 100 μg/mL∣2,4'-DDE 100 μg/mL∣2,4'-DDT 100 μg/mL∣4,4'-DDD 100 μg/mL∣4,4'-DDE 100 μg/mL∣4,4'-DDT 100 μg/mL∣Aldrin 100 μg/mL∣alpha-Endosulfan 100 μg/mL∣alpha-HCH 100 μg/mL∣beta-Endosulfan 100 μg/mL∣beta-HCH 100 μg/mL∣Cis-Chlordane (Alpha Isomer) 100 μg/mL∣delta-HCH 100 μg/mL∣Dieldrin 100 μg/mL∣Endosulfan-sulfate 100 μg/mL∣Endrin 100 μg/mL∣gamma-HCH 100 μg/mL∣Heptachlor 100 μg/mL∣Heptachlor-endo-epoxide (trans-, isomer A) 100 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 100 μg/mL∣Hexachlorobenzene 100 μg/mL∣Methoxychlor 100 μg/mL∣Mirex 100 μg/mL∣oxy-Chlordane 100 μg/mL∣Quintozene 100 μg/mL∣Trans-Chlordane (Gamma Isomer) 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?