missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PES1 Polyclonal Antibody

Description
Immunogen sequence: LYQLLNLHYP PKLEGQAQAE AKAGEGTYAL DSESCMEKLA ALSASLARVV VPATEEEAEV DEFPTDGEMS AQEEDRRKE Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 84%.
Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.
Specifications
Specifications
| Antigen | PES1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Pes1 |
| Gene Accession No. | O00541 |
| Gene Alias | HAMAP-Rule:MF_03028}; Pes; PES1; Pescadillo homolog; pescadillo homolog {ECO:0000255; pescadillo homolog 1, containing BRCT domain; pescadillo homolog 1, containing BRCT domain (zebrafish); pescadillo ribosomal biogenesis factor 1 |
| Gene Symbols | Pes1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?