missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pertussis Toxin

Product Code. 10151113
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10151113 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10151113 Supplier Invitrogen™ Supplier No. PHZ1174

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Formulation: Protein expressed in E. coli. Purity: >95%. Format: Lyophilized.

Islet-activating protein Pertussis toxin consists of an A protomer subunit (S1) which possesses both NAD+ glycohydrolase and ADP-ribosyltransferase activities, and B oligomer subunits (S2, S3, S4, and S5) which are responsible for attachment of the native toxin to eukaryotic cell surfaces. Pertussis toxin uncouples G proteins from receptors by ADP ribosylating a cysteine residue near the carboxyl terminus of the α subunit.

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 2665068
Purification Method PAGE
Quantity 50 μg
Content And Storage Store in refrigerator (2–8°C).
Inhibitor Properties Protein
Inhibitors GTPase Inhibitor
Shipping Condition Wet Ice
Specific Inhibit or Target G Proteins
Form Lyophilized
Purity or Quality Grade >95%
compliance-icons
Product Identifier
  • Pertussis Toxin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P264-Wash hands thoroughly after handling.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contain: Pertussis toxin

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.