missing translation for 'onlineSavingsMsg'
Learn More
Learn More
persephin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19713-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to PSPN.
Exhibits neurotrophic activity on mesencephalic dopaminergic and motor neurons.Specifications
| persephin | |
| Polyclonal | |
| Unconjugated | |
| PSPN | |
| persephin, PSP, PSPN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5623 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60542 | |
| PSPN | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction