missing translation for 'onlineSavingsMsg'
Learn More

Persephin Antibody (275508) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30736959 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736959 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736959 Supplier Novus Biologicals Supplier No. FAB2388JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Persephin Monoclonal antibody specifically detects Persephin in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Persephin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 275508
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias persephin, PSP
Host Species Mouse
Immunogen E. coli-derived recombinant human Persephin, Ala61-Gly156, Accession # O60542
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 5623
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.