missing translation for 'onlineSavingsMsg'
Learn More

Perrhenic acid, 75-80% aq. soln., Thermo Scientific Chemicals

Catalog Number 11351229
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
13768-11-1
HO4Re
251.21
MFCD00011326
UGSFIVDHFJJCBJ-UHFFFAOYSA-M
perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
83718
hydroxy(trioxo)rhenium
O[Re](=O)(=O)=O
This item is not returnable. View return policy
13768-11-1
HO4Re
251.21
MFCD00011326
UGSFIVDHFJJCBJ-UHFFFAOYSA-M
perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
83718
hydroxy(trioxo)rhenium
O[Re](=O)(=O)=O

Perrhenic acid is used in the manufacturing of superconducting synthetic metals and X-ray targets. It is used as a hydrocracking catalyst in petroleum industry and used for the dehydrogenation process and epoxidation of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless through Yellow
Physical Form Liquid
Quantity 25 g
Assay Percent Range 75-80%
Solubility Information Soluble in water.
Formula Weight 251.26
Concentration or Composition (by Analyte or Components) Perrhenic acid: 80.0%; Water: 20.0%
Appearance Colorless to pale yellow liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 2.42
Chemical Name or Material Perrhenic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'