missing translation for 'onlineSavingsMsg'
Learn More

Perphenazine Sulphoxide 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30464205
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464205 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464205 Supplier LoGiCal Supplier No. LGCAMP0826.0101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perphenazine Sulfoxide
CAS 10078-25-8
Analyte or Component Names Perphenazine Sulfoxide
Shipping Condition Room Temperature
Molecular Formula C21 H26 Cl N3 O2 S
Certifications/Compliance ISO 17034
Synonym Perphenazine Imp. A (EP), Perphenazine Sulfoxide, 2-[4-[3-(2-Chloro-5-oxido-10H-phenothiazin-10-yl)propyl]piperazin-1-yl]ethanol, Perphenazine Sulphoxide, Perphenazine Impurity A
InChI Formula InChI=1S/C21H26ClN3O2S/c22-17-6-7-21-19(16-17)25(18-4-1-2-5-20(18)28(21)27)9-3-8-23-10-12-24(13-11-23)14-15-26/h1-2,4-7,16,26H,3,8-15H2
Solution Type Single Solution
SMILES OCCN1CCN(CCCN2c3ccccc3S(=O)c4ccc(Cl)cc24)CC1
IUPAC Name 2-[4-[3-(2-chloro-5-oxophenothiazin-10-yl)propyl]piperazin-1-yl]ethanol
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 419.97
Quantity 1 mL
Formula Weight 419.1434 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.