missing translation for 'onlineSavingsMsg'
Learn More

Perphenazine Sulfoxide, TRC

Product Code. 30411889
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411889 250 mg -
30425608 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30411889 Supplier Toronto Research Chemicals Supplier No. TRCP291105250MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10078-25-8
Molecular Formula C21 H26 Cl N3 O2 S
Molecular Weight (g/mol) 419.97
Synonym 2-[4-[3-(2-Chloro-5-oxido-10H-phenothiazin-10-yl)propyl]piperazin-1-yl]ethanol, Perphenazine Imp. A (EP), Perphenazine Sulphoxide
IUPAC Name 2-[4-[3-(2-chloro-5-oxophenothiazin-10-yl)propyl]piperazin-1-yl]ethanol
SMILES OCCN1CCN(CCCN2c3ccccc3S(=O)c4ccc(Cl)cc24)CC1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H26ClN3O2S/c22-17-6-7-21-19(16-17)25(18-4-1-2-5-20(18)28(21)27)9-3-8-23-10-12-24(13-11-23)14-15-26/h1-2,4-7,16,26H,3,8-15H2
Formula Weight 419.1434
Percent Purity >95%
Chemical Name or Material Perphenazine Sulfoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.