missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PERP Recombinant Rabbit Monoclonal Antibody (1O3L5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538148
This item is not returnable.
View return policy
Description
Positive Samples: HepG2, BxPC-3, A-431, Mouse liver, Mouse kidney, Rat lung, Rat liver Immunogen sequence: LSFFALCGPQ MLVFLRVIGG LLALAAVFQI ISLVIYPVKY TQTFTLHANP AVTYIYNWAY GFGWAATIIL IGCAFFFCCL PNYEDDLLGN AKPRYFYTSA.
The p53 tumor-suppressor gene integrates numerous signals that control cell life and death. Several novel molecules involved in p53 network, including Chk2, p53R2, p53AIP1, Noxa, PIDD, PID/MTA2, MTBP and PERP, were identified and their genes were cloned recently. PERP, also termed PIGPC1 and THW, is a plasma membrane protein. p53 binds to the promoter of PERP and transcriptionally activates PERP gene then the translated PERP protein mediates the p53 induced apoptosis. The expression of PERP causes cell death. PERP is a mediator of p53 induced apoptosis. PERP has sequence similarity to PMP-22/gas3 and is a new member of the PMP-22/gas3 family.
Specifications
| PERP | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96FX8, Q9JK95 | |
| PERP | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1O3L5 | |
| Unconjugated | |
| PERP | |
| 1110017A08Rik; dJ496H19.1; HGNC:17637; KCP1; KCP-1; keratinocyte associated protein 1; keratinocyte-associated protein 1; keratinocytes associated protein 1; KRTCAP1; p53 apoptosis effector related to PMP22; p53 apoptosis effector related to PMP-22; p53 apoptosis-associated target; p53-induced protein PIGPC1; PERP; PERP, TP53 apoptosis effector; PIGPC1; RP3-496H19.1; THW; Transmembrane protein THW | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 292949, 64058, 64065 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction