missing translation for 'onlineSavingsMsg'
Learn More

PERP Rabbit anti-Human, Mouse, Rat, Clone: 1O3L5, Novus Biologicals™

Product Code. 18330877 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330877 20 μg 20µL
18335032 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18330877 Supplier Novus Biologicals Supplier No. NBP31661720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PERP Monoclonal antibody specifically detects PERP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PERP
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1O3L5
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias dJ496H19.1, KCP1KCP-1, Keratinocyte-associated protein 1, KRTCAP11110017A08Rik, p53 apoptosis effector related to PMP22, p53 apoptosis effector related to PMP-22, P53-induced protein PIGPC1, PERP, TP53 apoptosis effector, PIGPC1RP3-496H19.1, THWkeratinocytes associated protein 1, Transmembrane protein THW
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 94-193 of human PERP (Q96FX8).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 64065
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.