missing translation for 'onlineSavingsMsg'
Learn More

PERP Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18414668 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414668 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414668 Supplier Novus Biologicals Supplier No. NBP176741AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PERP Polyclonal antibody specifically detects PERP in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PERP
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias dJ496H19.1, KCP1KCP-1, Keratinocyte-associated protein 1, KRTCAP11110017A08Rik, p53 apoptosis effector related to PMP22, p53 apoptosis effector related to PMP-22, P53-induced protein PIGPC1, PERP, TP53 apoptosis effector, PIGPC1RP3-496H19.1, THWkeratinocytes associated protein 1, Transmembrane protein THW
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human PERP. The immunogen is located within the last 50 amino acids of PERP. Amino Acid Sequence: EDDLLGNAKPRYFYTSA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 64065
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.