missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Peroxy Orange 1

Product Code. 30001561
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001561 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001561 Supplier Tocris Bioscience™ Supplier No. 4944/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescent hydrogen peroxide indicator

Fluorescent probe for imaging hydrogen peroxide (H2O2); cell permeable. Displays an orange intracellular fluorescence in response to H2O2 signals produced in RAW 264.7 macrophages and A431 cells during immune response and growth factor stimulation, respectively. Excitation wavelength 543 nm, emission between 545 and 750 nm.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1199576-10-7
Quantity 10 mg
Molecular Formula C32H32BNO5
Formula Weight 521.41
Purity >98%
Molecular Weight (g/mol) 521.42
Chemical Name or Material Peroxy Orange 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.