missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin 6 Mouse anti-Human, Mouse, Clone: 2.20E+08, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59436-50UL
This item is not returnable.
View return policy
Description
Peroxiredoxin 6 Monoclonal specifically detects Peroxiredoxin 6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Peroxiredoxin 6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| 1-Cys, 24 kDa protein, Acidic calcium-independent phospholipase A2, aiPLA21-Cys PRX, Antioxidant protein 2, AOP2Non-selenium glutathione peroxidase, EC 1.11.1.15, EC 1.11.1.7, EC 3.1.1.-, KIAA0106Red blood cells page spot 12, Liver 2D page spot 40, MGC46173, NSGPx1-Cys peroxiredoxin, p29, peroxiredoxin 6, peroxiredoxin-6, PRX | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 9588 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 220000000 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| PRDX6 | |
| Recombinant human PRDX6 (1-224aa) purified from E. coli | |
| 50 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction