missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin 6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55159
This item is not returnable.
View return policy
Description
Peroxiredoxin 6 Polyclonal specifically detects Peroxiredoxin 6 in Human, Rat, Porcine samples. It is validated for Western Blot.
Specifications
| Peroxiredoxin 6 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| 1-Cys, 24 kDa protein, Acidic calcium-independent phospholipase A2, aiPLA21-Cys PRX, Antioxidant protein 2, AOP2Non-selenium glutathione peroxidase, EC 1.11.1.15, EC 1.11.1.7, EC 3.1.1.-, KIAA0106Red blood cells page spot 12, Liver 2D page spot 40, MGC46173, NSGPx1-Cys peroxiredoxin, p29, peroxiredoxin 6, peroxiredoxin-6, PRX | |
| Rabbit | |
| 25 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P30041 | |
| PRDX6 | |
| Synthetic peptides corresponding to PRDX6(peroxiredoxin 6) The peptide sequence was selected from the middle region of PRDX6 (NP_004896). Peptide sequence ARVVFVFGPDKKLKLSILYPATTGRNFDEILRVVISLQLTAEKRVATPVD. | |
| Protein A purified | |
| RUO | |
| 9588 | |
| Human, Rat, Pig, Bovine, Canine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction