missing translation for 'onlineSavingsMsg'
Learn More

Peroxiredoxin 6 Antibody, Novus Biologicals™

Product Code. 18149711 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18149711 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18149711 Supplier Novus Biologicals Supplier No. NBP219779

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peroxiredoxin 6 Polyclonal antibody specifically detects Peroxiredoxin 6 in Human, Mouse, Rat, Fish samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peroxiredoxin 6
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.89mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P30041
Gene Alias 1-Cys, 24 kDa protein, Acidic calcium-independent phospholipase A2, aiPLA21-Cys PRX, Antioxidant protein 2, AOP2Non-selenium glutathione peroxidase, EC 1.11.1.15, EC 1.11.1.7, EC 3.1.1.-, KIAA0106Red blood cells page spot 12, Liver 2D page spot 40, MGC46173, NSGPx1-Cys peroxiredoxin, p29, peroxiredoxin 6, peroxiredoxin-6, PRX
Gene Symbols PRDX6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Peroxiredoxin 6. The exact sequence is proprietary.
Molecular Weight of Antigen 25 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9588
Test Specificity Human, Mouse, Rat. Species reactivity based on sequence identity: Chicken (82%), Pig (90%), Rat (88%), Bovine (94%).
Target Species Human, Mouse, Rat, Fish
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.