missing translation for 'onlineSavingsMsg'
Learn More

Peroxiredoxin 6 Antibody (3A10-2A11), Novus Biologicals™

Product Code. 18356238 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356238 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356238 Supplier Novus Biologicals Supplier No. H00009588M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peroxiredoxin 6 Monoclonal antibody specifically detects Peroxiredoxin 6 in Human, Bovine samples. It is validated for Western Blot, ELISA, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peroxiredoxin 6
Applications Western Blot, ELISA, Dot Blot
Classification Monoclonal
Clone 3A10-2A11
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH35857
Gene Alias 1-Cys, 24 kDa protein, Acidic calcium-independent phospholipase A2, aiPLA21-Cys PRX, Antioxidant protein 2, AOP2Non-selenium glutathione peroxidase, EC 1.11.1.15, EC 1.11.1.7, EC 3.1.1.-, KIAA0106Red blood cells page spot 12, Liver 2D page spot 40, MGC46173, NSGPx1-Cys peroxiredoxin, p29, peroxiredoxin 6, peroxiredoxin-6, PRX
Host Species Mouse
Immunogen PRDX6 (AAH35857.1, 1 a.a. ~ 224 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPGGLLLGDVAPNFEANTTVGRIRFHDFLGDSWGILFSHPRDFTPVCTTELGRAAKLAPEFAKRNVKLIALSIDSVEDHLAWSKDINAYNCEEPTEKLPFPIIDDRNRELAILLGMLDPAEKDEKGMPVTARVVFVFGPDKKLKLSILYPATTGRNFDEILRVVISLQLTAEKRVATPVDWKDGDSVMVLPTIPEEEAKKLFPKGVFTKELPSGKKYLRYTPQP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9588
Target Species Human, Bovine
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.