missing translation for 'onlineSavingsMsg'
Learn More

Peroxiredoxin 6 Antibody (22E7), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18412873 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412873 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412873 Supplier Novus Biologicals Supplier No. NBP259436MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peroxiredoxin 6 Monoclonal antibody specifically detects Peroxiredoxin 6 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peroxiredoxin 6
Applications Flow Cytometry
Classification Monoclonal
Clone 220000000
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias 1-Cys, 24 kDa protein, Acidic calcium-independent phospholipase A2, aiPLA21-Cys PRX, Antioxidant protein 2, AOP2Non-selenium glutathione peroxidase, EC 1.11.1.15, EC 1.11.1.7, EC 3.1.1.-, KIAA0106Red blood cells page spot 12, Liver 2D page spot 40, MGC46173, NSGPx1-Cys peroxiredoxin, p29, peroxiredoxin 6, peroxiredoxin-6, PRX
Host Species Mouse
Immunogen Recombinant human Peroxiredoxin 6 (1-224aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9588
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.