missing translation for 'onlineSavingsMsg'
Learn More

Peroxiredoxin 5 Mouse anti-Human, Clone: 6A10, Novus Biologicals™

Mouse Monoclonal Antibody

£234.00 - £378.00

Specifications

Antigen Peroxiredoxin 5
Clone 6A10
Concentration 1 mg/mL
Dilution Western Blot 1:5000, Flow Cytometry 1:10-1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18244923
View Documents
Novus Biologicals
NBP2-59388-50UL
50 μL
£234.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18234113
View Documents
Novus Biologicals
NBP2-59388-100UL
100 μL
£378.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Peroxiredoxin 5 Monoclonal specifically detects Peroxiredoxin 5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Peroxiredoxin 5
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Human
ACR1Peroxisomal antioxidant enzyme, Alu corepressor 1, Alu co-repressor 1, Antioxidant enzyme B166, AOEB166Peroxiredoxin V, B166, EC 1.11.1.15, Liver tissue 2D-page spot 71B, MGC117264, MGC142283, MGC142285, peroxiredoxin 5, peroxiredoxin-5, mitochondrial, PLPThioredoxin peroxidase PMP20, PMP20, PRDX6, prx-V, PRXV, SBBI10, Thioredoxin reductase, TPx type VI
PRDX5
IgG1 κ
Protein A purified
6A10
Western Blot 1:5000, Flow Cytometry 1:10-1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Monoclonal
Purified
RUO
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
25824
Recombinant human Peroxiredoxin 5 (53-214aa) purified from E. coli
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.