missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin 5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38370-25ul
This item is not returnable.
View return policy
Description
Peroxiredoxin 5 Polyclonal specifically detects Peroxiredoxin 5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Peroxiredoxin 5 | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| P30044 | |
| PRDX5 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MGLAGVCALRRSAGYILVGGAGGQSAAAAARRCSEGEWASGGVRSFSRAAAAMAPIKVGDAIPAVEVFEGEPGN | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ACR1Peroxisomal antioxidant enzyme, Alu corepressor 1, Alu co-repressor 1, Antioxidant enzyme B166, AOEB166Peroxiredoxin V, B166, EC 1.11.1.15, Liver tissue 2D-page spot 71B, MGC117264, MGC142283, MGC142285, peroxiredoxin 5, peroxiredoxin-5, mitochondrial, PLPThioredoxin peroxidase PMP20, PMP20, PRDX6, prx-V, PRXV, SBBI10, Thioredoxin reductase, TPx type VI | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 25824 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction