missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peroxiredoxin 5 Antibody (125), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89863CL1
This item is not returnable.
View return policy
Description
Peroxiredoxin 5 Monoclonal antibody specifically detects Peroxiredoxin 5 in Human samples. It is validated for ELISA
Specifications
| Peroxiredoxin 5 | |
| Monoclonal | |
| CoraFluor 1 | |
| ACR1Peroxisomal antioxidant enzyme, Alu corepressor 1, Alu co-repressor 1, Antioxidant enzyme B166, AOEB166Peroxiredoxin V, B166, EC 1.11.1.15, Liver tissue 2D-page spot 71B, MGC117264, MGC142283, MGC142285, peroxiredoxin 5, peroxiredoxin-5, mitochondrial, PLPThioredoxin peroxidase PMP20, PMP20, PRDX6, prx-V, PRXV, SBBI10, Thioredoxin reductase, TPx type VI | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Peroxiredoxin 5 (Uniprot#: P30044-2; Met53-Leu214). | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cell Cycle and Replication | |
| 25824 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 125 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction