missing translation for 'onlineSavingsMsg'
Learn More

Peroxiredoxin 5 Antibody (125), Alexa Fluor™ 700 , Novus Biologicals™

Product Code. 18431809 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431809 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431809 Supplier Novus Biologicals Supplier No. NBP289863AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Peroxiredoxin 5 Monoclonal antibody specifically detects Peroxiredoxin 5 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peroxiredoxin 5
Applications ELISA
Classification Monoclonal
Clone 125
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias ACR1Peroxisomal antioxidant enzyme, Alu corepressor 1, Alu co-repressor 1, Antioxidant enzyme B166, AOEB166Peroxiredoxin V, B166, EC 1.11.1.15, Liver tissue 2D-page spot 71B, MGC117264, MGC142283, MGC142285, peroxiredoxin 5, peroxiredoxin-5, mitochondrial, PLPThioredoxin peroxidase PMP20, PMP20, PRDX6, prx-V, PRXV, SBBI10, Thioredoxin reductase, TPx type VI
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Peroxiredoxin 5 (P30044-2; Met53-Leu214).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 25824
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.