missing translation for 'onlineSavingsMsg'
Learn More

Peroxiredoxin 5 Antibody (002), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18341161 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341161 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341161 Supplier Novus Biologicals Supplier No. NBP289862JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Peroxiredoxin 5 Monoclonal specifically detects Peroxiredoxin 5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Knockout Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peroxiredoxin 5
Applications Immunocytochemistry, Immunofluorescence, Immunoassay
Classification Monoclonal
Clone 2
Conjugate Janelia Fluor 549
Dilution Immunocytochemistry/Immunofluorescence, Knockout Validated
Formulation 50 mM Sodium Borate
Gene Alias ACR1Peroxisomal antioxidant enzyme, Alu corepressor 1, Alu co-repressor 1, Antioxidant enzyme B166, AOEB166Peroxiredoxin V, B166, EC 1.11.1.15, Liver tissue 2D-page spot 71B, MGC117264, MGC142283, MGC142285, peroxiredoxin 5, peroxiredoxin-5, mitochondrial, PLPThioredoxin peroxidase PMP20, PMP20, PRDX6, prx-V, PRXV, SBBI10, Thioredoxin reductase, TPx type VI
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Peroxiredoxin 5 (P30044-2; Met53-Leu214).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25824
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.