missing translation for 'onlineSavingsMsg'
Learn More

Peroxiredoxin 4 Antibody, HRP, Novus Biologicals™

Product Code. 18320426 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320426 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320426 Supplier Novus Biologicals Supplier No. NBP299498H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peroxiredoxin 4 Polyclonal specifically detects Peroxiredoxin 4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peroxiredoxin 4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias Antioxidant enzyme AOE372, AOE37-2PRX-4, EC 1.11.1.15, peroxiredoxin 4, Peroxiredoxin IV, peroxiredoxin-4, prx4, prx-4, Prx-IV, thioredoxin peroxidase (antioxidant enzyme), Thioredoxin peroxidase AO372, Thioredoxin-dependent peroxide reductase A0372
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Peroxiredoxin 4.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10549
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.