missing translation for 'onlineSavingsMsg'
Learn More

Peroxiredoxin 3 Antibody [FITC], Novus Biologicals™

Product Code. 30735876 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735876 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735876 Supplier Novus Biologicals Supplier No. AF6610F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Peroxiredoxin 3 Polyclonal antibody specifically detects Peroxiredoxin 3 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peroxiredoxin 3
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias Antioxidant protein 1MGC104387, AOP-1MGC24293, AOP1PRO1748, EC 1.11.1.15, HBC189, MER5, peroxiredoxin 3, Peroxiredoxin III, peroxiredoxin-3, Protein MER5 homolog, prx-III, SP-22, thioredoxin-dependent peroxide reductase, mitochondrial
Host Species Goat
Immunogen E. coli-derived recombinant mouse Peroxiredoxin 3, Met1-Gln257, Accession # P20108
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10935
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.